Assay Details
Gene Symbol: Nrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146150.2 | NP_666262.2 | 4 - 5 | 933 | 107 | |
GenBank mRNA | AK031548.1 | - | 4 - 5 | 933 | 107 | |
AK040377.1 | - | 3 - 4 | 1164 | 107 | ||
AK166470.1 | - | 4 - 5 | 1063 | 107 | ||
BC023786.1 | - | 3 - 4 | 311 | 107 | ||
BC026832.1 | - | 4 - 5 | 983 | 4486176 | 107 | |
BC036128.1 | - | 4 - 5 | 1051 | 4971791 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
