Assay Details
Gene Symbol: Efcab10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029152.1 | NP_083428.1 | 1 - 2 | 341 | 103 | |
GenBank mRNA | AK015697.1 | - | 1 - 2 | 343 | 103 | |
BC099564.1 | - | 1 - 2 | 341 | 103 | ||
BC119387.1 | - | 1 - 2 | 236 | 103 | ||
BC120662.1 | - | 1 - 2 | 236 | 103 | ||
BF459438.1 | - | 3 - 4 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
