Assay Details
Gene Symbol: Efcab10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029152.1 | NP_083428.1 | 2 - 3 | 514 | 117 | |
GenBank mRNA | AK015697.1 | - | 2 - 3 | 516 | 117 | |
BC099564.1 | - | 2 - 3 | 514 | 117 | ||
BC119387.1 | - | 2 - 3 | 409 | 117 | ||
BC120662.1 | - | 2 - 3 | 409 | 117 | ||
BF459438.1 | - | 2 - 3 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
