Assay Details
Gene Symbol: Efcab10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029152.1 | NP_083428.1 | 3 - 4 | 601 | 117 | |
GenBank mRNA | AK015697.1 | - | 3 - 4 | 603 | 117 | |
BC099564.1 | - | 3 - 4 | 601 | 117 | ||
BC119387.1 | - | 3 - 4 | 496 | 117 | ||
BC120662.1 | - | 3 - 4 | 496 | 117 | ||
BF459438.1 | - | 1 - 2 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P467629 | BC099564.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P467629 | BF459438.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P467629 | NM_029152.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P467629 | AK015697.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P467629 | BC099564.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P467629 | BF459438.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P467629 | NM_029152.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P467629 | AK015697.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355362 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 521787 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 162462 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 950234 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730924 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52598 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138952 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P467629 | BC099564.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P467629 | BF459438.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P467629 | NM_029152.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P467629 | AK015697.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P467629 | BC099564.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P467629 | BF459438.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P467629 | NM_029152.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P467629 | AK015697.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138952 | - | - |
Whole Mouse Genome G4122A | A_51_P467629 | BC099564.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P467629 | BF459438.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P467629 | NM_029152.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P467629 | AK015697.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P467629 | BC099564.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P467629 | BF459438.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P467629 | NM_029152.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P467629 | AK015697.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138952 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341824 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341812 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341842 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P10282429 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341837 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341838 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341862 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341860 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341832 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P10352890 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09906696 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341813 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P10417779 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09825242 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P10258509 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341804 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341858 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341815 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341840 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341799 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P10164666 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341836 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P10022316 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341800 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341802 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09791142 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341798 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341819 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341793 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341807 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341814 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341796 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09825243 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341805 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P10095887 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341816 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28341822 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10395307 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10395311 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10395309 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10395310 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10395308 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1453558_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10395306 | - | - |
Mouse Genome 430 2.0 Array | 1453558_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1453558_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75040_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75040_at | - | - |
Mouse Expression Set 430 | 1453558_PM_at | - | - |
Mouse Expression Set 430 | 1453558_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453558_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453558_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 808610 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2725119 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2725119 | - | - |
