Assay Details
| Gene Symbol: Slc2a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019741.3 | NP_062715.2 | 6 - 7 | 1070 | 108 | |
| GenBank mRNA | AF161071.1 | - | 4 - 5 | 431 | 108 | |
| AK029720.1 | - | 6 - 7 | 1071 | 108 | ||
| BC023500.1 | - | 4 - 5 | 446 | 5043284 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc2a5
Genomic Map