Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800005049.mm.1 |
NM_025468.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005049.mm.1 |
NM_025468.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005049.mm.1 |
TC1800000682.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800005050.mm.1 |
NM_025468.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005050.mm.1 |
NM_025468.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005050.mm.1 |
TC1800000682.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800005054.mm.1 |
NM_025468.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005054.mm.1 |
NM_025468.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800005054.mm.1 |
TC1800000682.mm.1 |
MTA TC |