Assay Details
| Gene Symbol: Pcyox1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172832.4 | NP_766420.1 | 5 - 6 | 879 | 68 | |
| GenBank mRNA | AK050006.1 | - | 5 - 6 | 880 | 68 | |
| BC060677.1 | - | 5 - 6 | 869 | 6404883 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcyox1l
Genomic Map