Assay Details
| Gene Symbol: Htr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008313.4 | NP_032339.2 | 4 - 5 | 565 | 64 | |
| GenBank mRNA | AJ011369.1 | - | 3 - 4 | 355 | 64 | |
| AK141587.1 | - | 4 - 5 | 509 | 64 | ||
| AK141625.1 | - | 4 - 5 | 566 | 64 | ||
| BC132189.1 | - | 3 - 4 | 394 | 64 | ||
| BC132191.1 | - | 3 - 4 | 394 | 64 | ||
| BC145159.1 | - | 3 - 4 | 394 | 64 | ||
| BC145161.1 | - | 3 - 4 | 394 | 64 | ||
| JN418927.1 | - | 4 - 5 | 496 | 64 | ||
| JN418928.1 | - | 4 - 5 | 482 | 64 | ||
| JN418929.1 | - | 3 - 4 | 355 | 64 | ||
| JN418930.1 | - | 3 - 4 | 371 | 64 | ||
| Y09585.1 | - | 3 - 4 | 355 | 64 | ||
| Y09587.1 | - | 3 - 4 | 355 | 64 | ||
| Y09588.1 | - | 3 - 4 | 355 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Htr4
Genomic Map