Assay Details
| Gene Symbol: Csnk1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146087.2 | NP_666199.1 | 3 - 4 | 520 | 141 | |
| GenBank mRNA | AK076070.1 | - | 3 - 4 | 787 | 141 | |
| AK076264.1 | - | 3 - 4 | 781 | 141 | ||
| AK078398.1 | - | 3 - 4 | 769 | 141 | ||
| AK141421.1 | - | 3 - 4 | 774 | 142 | ||
| AK146873.1 | - | 3 - 4 | 790 | 141 | ||
| AK147165.1 | - | 3 - 4 | 765 | 141 | ||
| AK152387.1 | - | 3 - 4 | 765 | 141 | ||
| AK152793.1 | - | 3 - 4 | 785 | 141 | ||
| AK166318.1 | - | 3 - 4 | 780 | 141 | ||
| AK166429.1 | - | 3 - 4 | 762 | 141 | ||
| AK167550.1 | - | 3 - 4 | 788 | 141 | ||
| BC019740.1 | - | 3 - 4 | 520 | 3487913 | 141 | |
| BC025439.1 | - | 3 - 4 | 467 | 5038433 | 141 | |
| BC048081.1 | - | 3 - 4 | 553 | 6309504 | 141 | |
More Information
Updated Mapping Information
Set Membership: |
|

Csnk1a1
Genomic Map