Assay Details
| Gene Symbol: Csnk1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146087.2 | NP_666199.1 | 5 - 6 | 769 | 100 | |
| GenBank mRNA | AK076070.1 | - | 5 - 6 | 1036 | 100 | |
| AK078398.1 | - | 5 - 6 | 1018 | 100 | ||
| AK141421.1 | - | 5 - 6 | 1024 | 100 | ||
| AK146873.1 | - | 5 - 6 | 1039 | 100 | ||
| AK147165.1 | - | 5 - 6 | 1014 | 100 | ||
| AK152387.1 | - | 5 - 6 | 1014 | 100 | ||
| AK152793.1 | - | 5 - 6 | 1034 | 100 | ||
| AK159939.1 | - | 2 - 3 | 238 | 100 | ||
| AK167550.1 | - | 5 - 6 | 1037 | 100 | ||
| AK180731.1 | - | 2 - 3 | 212 | 100 | ||
| AK219793.1 | - | 2 - 3 | 180 | 100 | ||
| BC019740.1 | - | 5 - 6 | 769 | 3487913 | 100 | |
| BC048081.1 | - | 5 - 6 | 802 | 6309504 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Csnk1a1
Genomic Map