Assay Details
| Gene Symbol: Csnk1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146087.2 | NP_666199.1 | 8 - 9 | 1027 | 99 | |
| GenBank mRNA | AK076070.1 | - | 8 - 9 | 1294 | 99 | |
| AK076264.1 | - | 8 - 9 | 1286 | 99 | ||
| AK078398.1 | - | 8 - 9 | 1276 | 99 | ||
| AK141421.1 | - | 8 - 9 | 1282 | 99 | ||
| AK146873.1 | - | 8 - 9 | 1297 | 99 | ||
| AK147165.1 | - | 8 - 9 | 1272 | 99 | ||
| AK152387.1 | - | 8 - 9 | 1272 | 99 | ||
| AK152793.1 | - | 8 - 9 | 1292 | 99 | ||
| AK159939.1 | - | 5 - 6 | 496 | 99 | ||
| AK166318.1 | - | 8 - 9 | 1287 | 99 | ||
| AK166429.1 | - | 8 - 9 | 1269 | 99 | ||
| AK167550.1 | - | 8 - 9 | 1295 | 99 | ||
| BC002171.1 | - | 2 - 3 | 175 | 99 | ||
| BC019740.1 | - | 8 - 9 | 1027 | 3487913 | 99 | |
| BC025439.1 | - | 8 - 9 | 974 | 5038433 | 99 | |
| BC048081.1 | - | 8 - 9 | 1060 | 6309504 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Csnk1a1
Genomic Map