Assay Details
| Gene Symbol: Casz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159344.1 | NP_001152816.1 | 14 - 15 | 3391 | 60 | |
| NM_027195.2 | NP_081471.2 | 14 - 15 | 3391 | 60 | ||
| GenBank mRNA | AK010559.1 | - | 3 - 4 | 301 | 60 | |
| AK173341.1 | - | 9 - 10 | 2181 | 60 | ||
| BC075673.1 | - | 14 - 15 | 3393 | 60 | ||
| BI738136.1 | - | 2 - 3 | 228 | 60 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Casz1
Genomic Map