Assay Details
| Gene Symbol: Plcd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081456.1 | NP_001074925.1 | 7 - 8 | 1146 | 92 | |
| NM_148937.2 | NP_683739.2 | 7 - 8 | 1297 | 92 | ||
| GenBank mRNA | AK016945.1 | - | 7 - 8 | 1294 | 92 | |
| AK029561.1 | - | 7 - 8 | 1292 | 92 | ||
| AK039149.1 | - | 7 - 8 | 1124 | 92 | ||
| AK140231.1 | - | 7 - 8 | 1133 | 92 | ||
| AY033991.1 | - | 7 - 8 | 1129 | 92 | ||
| BC066156.1 | - | 7 - 8 | 1037 | 30099811 | 92 | |
More Information
Set Membership: |
|

Plcd4
Genomic Map