Assay Details
| Gene Symbol: Abcb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023732.2 | NP_076221.1 | 2 - 3 | 857 | 75 | |
| GenBank mRNA | AF045562.1 | - | 2 - 3 | 719 | 75 | |
| AK004605.1 | - | 2 - 3 | 937 | 75 | ||
| AK040323.1 | - | 2 - 3 | 861 | 75 | ||
| AK150853.1 | - | 2 - 3 | 945 | 75 | ||
| AK151165.1 | - | 2 - 3 | 966 | 75 | ||
| AK152584.1 | - | 2 - 3 | 945 | 75 | ||
| AK164003.1 | - | 2 - 3 | 861 | 75 | ||
| AK168642.1 | - | 2 - 3 | 958 | 75 | ||
| BC006634.1 | - | 2 - 3 | 857 | 3710552 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF045562.1 transcripts.
Set Membership: |
|

Abcb6
Genomic Map