Assay Details
| Gene Symbol: Glb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029010.1 | NP_083286.1 | 2 - 3 | 312 | 62 | |
| GenBank mRNA | AK014667.1 | - | 1 - 2 | 117 | 62 | |
| AK029925.1 | - | 2 - 3 | 312 | 62 | ||
| AK030992.1 | - | 1 - 2 | 1375 | 62 | ||
| BC021773.1 | - | 1 - 2 | 65 | 4222994 | 62 | |
| BY275381.1 | - | 2 - 3 | 295 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014667.1 transcripts.
Set Membership: |
|

Glb1l
Genomic Map