Assay Details
| Gene Symbol: Glb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029010.1 | NP_083286.1 | 3 - 4 | 482 | 77 | |
| GenBank mRNA | AK029925.1 | - | 3 - 4 | 482 | 77 | |
| BC021773.1 | - | 2 - 3 | 235 | 4222994 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Glb1l
Genomic Map