Assay Details
Gene Symbol: Glb1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029010.1 | NP_083286.1 | 5 - 6 | 691 | 81 | |
GenBank mRNA | AK014667.1 | - | 3 - 4 | 714 | 81 | |
AK029925.1 | - | 5 - 6 | 691 | 81 | ||
BC021773.1 | - | 4 - 5 | 444 | 4222994 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
