Assay Details
| Gene Symbol: Glb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029010.1 | NP_083286.1 | 6 - 7 | 785 | 100 | |
| GenBank mRNA | AK014667.1 | - | 4 - 5 | 808 | 100 | |
| AK029925.1 | - | 6 - 7 | 785 | 100 | ||
| BC021773.1 | - | 5 - 6 | 538 | 4222994 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Glb1l
Genomic Map