Assay Details
| Gene Symbol: Glb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029010.1 | NP_083286.1 | 9 - 10 | 1135 | 89 | |
| GenBank mRNA | AK014667.1 | - | 7 - 8 | 1158 | 89 | |
| AK029925.1 | - | 9 - 10 | 1135 | 89 | ||
| BC021773.1 | - | 8 - 9 | 888 | 4222994 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Glb1l
Genomic Map