Assay Details
| Gene Symbol: Ttll4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014974.1 | NP_001014974.1 | 6 - 7 | 2174 | 84 | |
| GenBank mRNA | AK129075.1 | - | 4 - 5 | 1001 | 84 | |
| AM690747.1 | - | 4 - 5 | 1777 | 84 | ||
| BC085151.1 | - | 6 - 7 | 2174 | 30534216 | 84 | |
More Information
Set Membership: |
|

Ttll4
Genomic Map