Assay Details
| Gene Symbol: Slc23a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194333.3 | NP_919314.2 | 10 - 11 | 1481 | 101 | |
| GenBank mRNA | BC094893.1 | - | 10 - 11 | 1484 | 30017276 | 101 |
| BC117790.1 | - | 10 - 11 | 1468 | 40092152 | 101 | |
| BC117791.1 | - | 10 - 11 | 1468 | 40092153 | 101 | |
| U25739.1 | - | 10 - 11 | 1493 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1964752 | U25739.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1964752 | BC094893.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1964752 | NM_194333.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 890374 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 756998 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284848 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581213 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76709 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128598 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1964752 | U25739.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1964752 | BC094893.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1964752 | NM_194333.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P424550 | U25739.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P424550 | BC094893.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P424550 | NM_194333.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20352185 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128598 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04149529 | - | - |
| Whole Mouse Genome G4122A | A_51_P424550 | U25739.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P424550 | BC094893.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P424550 | NM_194333.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121381 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116717 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128598 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00958015 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01100956 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01100955 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00958010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20352227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01100955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00086696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00086697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352182 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01082025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00961111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00958016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00961113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01082028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00330480 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01082014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01082027 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00536618 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00086698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20352185 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10355726 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355727 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355721 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355729 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355725 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355718 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355720 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355719 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355724 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355722 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355723 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10355728 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460042_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10355717 | - | - |
| Mouse Genome 430 2.0 Array | 1460042_at | - | - |
| Mouse Genome 430 2.0 Array | 1460042_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22626_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22626_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100021053.mm.1 | NM_194333.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100021053.mm.1 | NM_194333.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100021053.mm.1 | TC0100002599.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1460042_at | - | - |
| Mouse Expression Set 430 | 1460042_PM_at | - | - |
| Murine Genome U74v2 Set | 98800_at | - | - |
| Murine Genome U74v2 Set | 161149_r_at | - | - |
| Murine Genome U74v2 Set | 98800_PM_at | - | - |
| Murine Genome U74v2 Set | 161149_PM_r_at | - | - |
| Murine Genome U74v2 Set | 169606_PM_r_at | - | - |
| Murine Genome U74v2 Set | 169606_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460042_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460042_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460042_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460042_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 908016 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6840152 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6840152 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006840152 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1218441 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1218441 | - | - |
| mouse-6 v1.1 (Illumina) | scl022626.1_322-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6840152 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl022626.1_322-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6840152 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1218441 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1218441 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl022626.1_322-S | - | - |

Slc23a3
Genomic Map