Assay Details
| Gene Symbol: Slc23a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194333.3 | NP_919314.2 | 5 - 6 | 725 | 69 | |
| GenBank mRNA | BC094893.1 | - | 5 - 6 | 729 | 30017276 | 69 |
| BC117790.1 | - | 5 - 6 | 712 | 40092152 | 69 | |
| BC117791.1 | - | 5 - 6 | 712 | 40092153 | 69 | |
| U25739.1 | - | 5 - 6 | 737 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc23a3
Genomic Map