Assay Details
| Gene Symbol: Slc23a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194333.3 | NP_919314.2 | 7 - 8 | 954 | 59 | |
| GenBank mRNA | BC094893.1 | - | 7 - 8 | 958 | 30017276 | 59 |
| BC117790.1 | - | 7 - 8 | 941 | 40092152 | 59 | |
| BC117791.1 | - | 7 - 8 | 941 | 40092153 | 59 | |
| U25739.1 | - | 7 - 8 | 966 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc23a3
Genomic Map