Assay Details
| Gene Symbol: Bcs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012324.1 | - | 6 - 7 | 1075 | 67 | |
| AK078925.1 | - | 6 - 7 | 1087 | 67 | ||
| AK079385.1 | - | 6 - 7 | 1083 | 67 | ||
| AK216891.1 | - | 1 - 2 | 90 | 67 | ||
| BC019781.1 | - | 6 - 7 | 975 | 4036047 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20349430 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096485 | AK212348.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096485 | AK079385.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096485 | NM_025784.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096486 | BC019781.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096486 | AK212348.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096486 | AK012324.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096486 | AK078925.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096486 | AK079385.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 124972 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 569974 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096485 | AK212348.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096485 | AK079385.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096485 | NM_025784.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096486 | BC019781.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096486 | AK212348.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096486 | AK012324.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096486 | AK078925.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096486 | AK079385.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104782 | BC019781.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104782 | AK212348.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104782 | AK012324.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104782 | AK078925.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P104782 | AK079385.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20349428 | - | - |
| Whole Mouse Genome G4122A | A_52_P104782 | BC019781.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104782 | AK212348.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104782 | AK012324.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104782 | AK078925.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P104782 | AK079385.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101017 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100961 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00776598 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00776600 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00592535 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20349430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20349503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20349451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20349446 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00776598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20349443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00463535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20349432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20349463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00592528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20349447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00592546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20349440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00592543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20349436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20349467 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20349430 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20349438 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00463538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20349460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20349458 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00592535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00592541 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00463541 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10347424 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347420 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347421 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347419 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347422 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347425 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347418 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347426 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10347423 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451541_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10347417 | - | - |
| Mouse Genome 430 2.0 Array | 1451541_at | - | - |
| Mouse Genome 430 2.0 Array | 1451541_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66821_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66821_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100005228.mm.1 | NM_025784.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100005228.mm.1 | TC0100000638.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0100005227.mm.1 | NM_025784.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100005227.mm.1 | TC0100000638.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451541_PM_at | - | - |
| Mouse Expression Set 430 | 1451541_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451541_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451541_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451541_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451541_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5360102 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005360102 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1248953 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1248953 | - | - |
| mouse-6 v1.1 (Illumina) | scl17795.8.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5360102 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl17795.8.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5360102 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2691345 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2691345 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl17795.8.1_1-S | - | - |

Bcs1l
Genomic Map