Assay Details
| Gene Symbol: Ifltd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028742.2 | NP_083018.1 | 2 - 3 | 149 | 147 | |
| GenBank mRNA | AK016641.1 | - | 2 - 3 | 149 | 147 | |
| BC053420.1 | - | 1 - 2 | 111 | 6705208 | 147 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK016641.1,BC053420.1,NM_028742.2 transcripts.
This assay is in the 5'UTR region of the AK016641.1,BC053420.1,NM_028742.2 transcripts.
Set Membership: |
|

Ifltd1
Genomic Map