Assay Details
Gene Symbol: Slc31a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175090.4 | NP_780299.2 | 2 - 3 | 333 | 133 | |
GenBank mRNA | AK046835.1 | - | 2 - 3 | 286 | 133 | |
AK151337.1 | - | 2 - 3 | 315 | 133 | ||
AK170285.1 | - | 2 - 3 | 286 | 133 | ||
BC034674.1 | - | 2 - 3 | 271 | 3968084 | 133 | |
BC058227.1 | - | 2 - 3 | 273 | 6309323 | 133 | |
BC065147.1 | - | 1 - 2 | 181 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
