Assay Details
| Gene Symbol: Zfp37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290350.1 | NP_001277279.1 | 2 - 3 | 314 | 77 | |
| NM_001290351.1 | NP_001277280.1 | 2 - 3 | 305 | 77 | ||
| NM_001290353.1 | NP_001277282.1 | 2 - 3 | 212 | 77 | ||
| NM_009554.4 | NP_033580.3 | 2 - 3 | 302 | 77 | ||
| GenBank mRNA | BC063757.1 | - | 2 - 3 | 186 | 6331110 | 77 |
| CJ078183.1 | - | 2 - 3 | 302 | 77 | ||
| CJ080611.1 | - | 2 - 3 | 302 | 77 | ||
| CO430987.1 | - | 2 - 3 | 184 | 77 | ||
| CO434318.1 | - | 2 - 3 | 139 | 77 | ||
| X89264.1 | - | 3 - 4 | 214 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001290353.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp37
Genomic Map