Assay Details
Gene Symbol: Ptprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207270.2 | NP_997153.2 | 10 - 11 | 2179 | 67 | |
GenBank mRNA | AB217856.1 | - | 9 - 10 | 1852 | 67 | |
AK078884.1 | - | 9 - 10 | 1856 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
