Assay Details
| Gene Symbol: 1700120E14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045368.1 | 2 - 3 | 796 | 89 | ||
| GenBank mRNA | AK007214.1 | - | 2 - 3 | 797 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myosin VB | Myo5b |
AI661750, mKIAA1119 |
Anti-sense | Chr. 18 - 74442378 - 74772698 on NCBI Build GRCm38 | |
| Downstream | small Cajal body-specific RNA 17 | Scarna17 | MBII-119 | 252.5kb | Sense | Chr. 18 - 74778394 - 74778455 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700120E14Rik
Genomic Map