Assay Details
| Gene Symbol: Eif4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159375.1 | NP_001152847.1 | 11 | 117 | ||
| NM_144958.4 | NP_659207.1 | 11 | 117 | |||
| GenBank mRNA | AK077429.1 | - | 11 | 117 | ||
| AK088124.1 | - | 7 | 117 | |||
| AK144850.1 | - | 11 | 117 | |||
| AK145375.1 | - | 11 | 117 | |||
| AK145508.1 | - | 11 | 117 | |||
| AK146424.1 | - | 11 | 1678 | 117 | ||
| AK146464.1 | - | 11 | 117 | |||
| AK152210.1 | - | 8 | 117 | |||
| AK152694.1 | - | 11 | 117 | |||
| AK159643.1 | - | 11 | 117 | |||
| AK166439.1 | - | 11 | 1658 | 117 | ||
| AK166527.1 | - | 11 | 117 | |||
| AK166572.1 | - | 11 | 117 | |||
| AK166674.1 | - | 11 | 117 | |||
| AK167003.1 | - | 11 | 117 | |||
| AK167083.1 | - | 11 | 117 | |||
| AK167196.1 | - | 11 | 117 | |||
| AK167205.1 | - | 11 | 117 | |||
| AK167906.1 | - | 10 | 117 | |||
| AK167988.1 | - | 11 | 117 | |||
| AK168697.1 | - | 11 | 117 | |||
| AK168780.1 | - | 11 | 117 | |||
| AK169023.1 | - | 11 | 117 | |||
| AK169030.1 | - | 11 | 117 | |||
| AK169057.1 | - | 11 | 117 | |||
| AK169156.1 | - | 11 | 117 | |||
| AK169279.1 | - | 11 | 117 | |||
| AK208666.1 | - | 1 | 266 | 117 | ||
| AW493973.1 | - | 1 | 117 | |||
| BC002266.1 | - | 2 | 644 | 117 | ||
| BC012282.1 | - | 11 | 1663 | 117 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eif4a1
Genomic Map