Assay Details
| Gene Symbol: Slc35d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK049462.1 | - | 10 - 11 | 807 | 118 | |
| AK169775.1 | - | 10 - 11 | 789 | 118 | ||
| AK220330.1 | - | 10 - 11 | 797 | 118 | ||
| BC115984.1 | - | 11 - 12 | 788 | 40105299 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc35d1
Genomic Map