Assay Details
Gene Symbol: Slc35d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK049462.1 | - | 11 - 12 | 896 | 155 | |
AK169775.1 | - | 11 - 12 | 878 | 155 | ||
AK220330.1 | - | 11 - 12 | 886 | 155 | ||
BC115984.1 | - | 12 - 13 | 877 | 40105299 | 155 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
