Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400007715.mm.1 |
NM_175471.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007715.mm.1 |
TC0400001120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007712.mm.1 |
NM_175471.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007712.mm.1 |
TC0400001120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007716.mm.1 |
NM_175471.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007716.mm.1 |
TC0400001120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007717.mm.1 |
NM_175471.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007717.mm.1 |
TC0400001120.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007713.mm.1 |
NM_175471.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007713.mm.1 |
TC0400001120.mm.1 |
MTA TC |