Assay Details
| Gene Symbol: Ssbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023672.2 | NP_076161.2 | 16 - 17 | 1426 | 69 | |
| NM_198438.1 | NP_940840.1 | 15 - 16 | 1345 | 69 | ||
| GenBank mRNA | AF170906.1 | - | 15 - 16 | 1075 | 69 | |
| AF500117.1 | - | 15 - 16 | 1075 | 69 | ||
| AK011509.1 | - | 16 - 17 | 1041 | 69 | ||
| AK011853.1 | - | 16 - 17 | 1396 | 69 | ||
| AK017711.1 | - | 16 - 17 | 1430 | 69 | ||
| AK041388.1 | - | 12 - 13 | 942 | 69 | ||
| AK153733.1 | - | 12 - 13 | 786 | 69 | ||
| AY167987.1 | - | 15 - 16 | 958 | 69 | ||
| BC003430.1 | - | 15 - 16 | 1099 | 2646149 | 69 | |
| BC035475.1 | - | 15 - 16 | 1288 | 69 | ||
More Information
Set Membership: |
|

Ssbp3
Genomic Map