Assay Details
| Gene Symbol: Ssbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023672.2 | NP_076161.2 | 11 - 12 | 1146 | 70 | |
| NM_198438.1 | NP_940840.1 | 10 - 11 | 1065 | 70 | ||
| GenBank mRNA | AF170906.1 | - | 10 - 11 | 795 | 70 | |
| AF500117.1 | - | 10 - 11 | 795 | 70 | ||
| AK011853.1 | - | 11 - 12 | 1116 | 70 | ||
| AK017711.1 | - | 11 - 12 | 1150 | 70 | ||
| AK041388.1 | - | 7 - 8 | 662 | 70 | ||
| AK153733.1 | - | 7 - 8 | 506 | 70 | ||
| AY167987.1 | - | 10 - 11 | 678 | 70 | ||
| BC003430.1 | - | 10 - 11 | 819 | 2646149 | 70 | |
| BC028324.1 | - | 5 - 6 | 287 | 70 | ||
| BC035475.1 | - | 10 - 11 | 1008 | 70 | ||
More Information
Set Membership: |
|

Ssbp3
Genomic Map