Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400016525.mm.1 |
NM_001146057.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016525.mm.1 |
NM_001146057.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016525.mm.1 |
NM_001146058.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016525.mm.1 |
NM_001146058.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016525.mm.1 |
NM_133348.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016525.mm.1 |
NM_133348.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016525.mm.1 |
TC0400002016.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400016526.mm.1 |
NM_001146057.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016526.mm.1 |
NM_001146057.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016526.mm.1 |
NM_001146058.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016526.mm.1 |
NM_001146058.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016526.mm.1 |
NM_133348.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016526.mm.1 |
NM_133348.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400016526.mm.1 |
TC0400002016.mm.1 |
MTA TC |