Assay Details
| Gene Symbol: Nr3c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008173.3 | NP_032199.3 | 5 - 6 | 1949 | 69 | |
| GenBank mRNA | AK037733.1 | - | 5 - 6 | 1830 | 69 | |
| AK155200.1 | - | 7 - 8 | 2031 | 69 | ||
| AK156323.1 | - | 7 - 8 | 2025 | 69 | ||
| BC129912.1 | - | 6 - 7 | 1927 | 40111802 | 69 | |
| BC129913.1 | - | 6 - 7 | 1927 | 40111803 | 69 | |
| DQ504162.1 | - | 5 - 6 | 1933 | 69 | ||
| HM236293.1 | - | 6 - 7 | 1909 | 69 | ||
| X04435.1 | - | 6 - 7 | 1927 | 69 | ||
| X13358.1 | - | 6 - 7 | 1909 | 69 | ||
| X13359.1 | - | 6 - 7 | 1912 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nr3c1
Genomic Map