Assay Details
| Gene Symbol: B3gntl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178664.5 | NP_848779.2 | 8 - 9 | 831 | 55 | |
| GenBank mRNA | AK031361.1 | - | 8 - 9 | 820 | 55 | |
| AK156318.1 | - | 8 - 9 | 809 | 55 | ||
| BC022739.1 | - | 2 - 3 | 143 | 55 | ||
| BC062106.1 | - | 2 - 3 | 690 | 55 | ||
| BC088993.1 | - | 8 - 9 | 696 | 6416623 | 55 | |
| BC125626.1 | - | 8 - 9 | 804 | 40130141 | 55 | |
| BC132193.1 | - | 8 - 9 | 804 | 40130470 | 55 | |
| BC145163.1 | - | 8 - 9 | 808 | 55 | ||
| BC145165.1 | - | 8 - 9 | 870 | 55 | ||
| BC145166.1 | - | 8 - 9 | 804 | 9053717 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

B3gntl1
Genomic Map