Assay Details
| Gene Symbol: Car6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009802.2 | NP_033932.2 | 1 - 2 | 96 | 84 | |
| GenBank mRNA | AF079835.1 | - | 1 - 2 | 123 | 84 | |
| AK132599.1 | - | 1 - 2 | 97 | 84 | ||
| AK161216.1 | - | 1 - 2 | 92 | 84 | ||
| BC006726.1 | - | 1 - 2 | 183 | 84 | ||
| BC018582.1 | - | 1 - 2 | 73 | 84 | ||
| BC036270.1 | - | 1 - 2 | 79 | 84 | ||
| BC039657.1 | - | 1 - 2 | 81 | 84 | ||
| BC049973.1 | - | 1 - 2 | 94 | 84 | ||
| BC055437.1 | - | 1 - 2 | 82 | 4190566 | 84 | |
More Information
Set Membership: |
|

Car6
Genomic Map