Assay Details
| Gene Symbol: Tle1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285529.1 | NP_001272458.1 | 3 - 4 | 1077 | 96 | |
| NM_001285530.1 | NP_001272459.1 | 3 - 4 | 1077 | 96 | ||
| NM_001285531.1 | NP_001272460.1 | 3 - 4 | 1077 | 96 | ||
| NM_001285532.1 | NP_001272461.1 | 3 - 4 | 1077 | 96 | ||
| NM_011599.5 | NP_035729.3 | 3 - 4 | 1077 | 96 | ||
| GenBank mRNA | AK044642.1 | - | 3 - 4 | 1079 | 96 | |
| AK046402.1 | - | 3 - 4 | 1045 | 96 | ||
| AK048515.1 | - | 3 - 4 | 1063 | 96 | ||
| AK052961.1 | - | 1 - 2 | 1178 | 96 | ||
| AK076750.1 | - | 3 - 4 | 459 | 96 | ||
| AK082499.1 | - | 3 - 4 | 1043 | 96 | ||
| AY155195.1 | - | 3 - 4 | 236 | 96 | ||
| AY155196.1 | - | 3 - 4 | 1046 | 96 | ||
| AY155197.1 | - | 3 - 4 | 235 | 96 | ||
| AY155199.1 | - | 3 - 4 | 236 | 96 | ||
| BC057573.1 | - | 3 - 4 | 907 | 6401650 | 96 | |
| U61362.1 | - | 3 - 4 | 243 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tle1
Genomic Map