Assay Details
| Gene Symbol: Rorc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK153941.1 | - | 2 - 3 | 233 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK153941.1 transcripts.
Set Membership: |
|

Rorc
Genomic Map