Assay Details
| Gene Symbol: Rorc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293734.1 | NP_001280663.1 | 2 - 3 | 256 | 95 | |
| NM_011281.3 | NP_035411.2 | 3 - 4 | 252 | 95 | ||
| NR_121656.1 | 2 - 3 | 256 | 95 | |||
| GenBank mRNA | AF163668.1 | - | 2 - 3 | 183 | 95 | |
| AJ132394.1 | - | 2 - 3 | 256 | 95 | ||
| AK137866.1 | - | 2 - 3 | 229 | 95 | ||
| AK153941.1 | - | 3 - 4 | 314 | 95 | ||
| AK160122.1 | - | 3 - 4 | 210 | 95 | ||
| BC014804.1 | - | 3 - 4 | 195 | 4489193 | 95 | |
| CJ098183.1 | - | 3 - 4 | 220 | 95 | ||
| U39071.1 | - | 3 - 4 | 228 | 95 | ||
| U43508.1 | - | 3 - 4 | 190 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rorc
Genomic Map