Assay Details
| Gene Symbol: Rorc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293734.1 | NP_001280663.1 | 4 - 5 | 905 | 73 | |
| NM_011281.3 | NP_035411.2 | 5 - 6 | 901 | 73 | ||
| NR_121656.1 | 4 - 5 | 905 | 73 | |||
| GenBank mRNA | AF163668.1 | - | 4 - 5 | 832 | 73 | |
| AJ132394.1 | - | 4 - 5 | 905 | 73 | ||
| AK137866.1 | - | 4 - 5 | 878 | 73 | ||
| AK153941.1 | - | 5 - 6 | 963 | 73 | ||
| AK160122.1 | - | 5 - 6 | 859 | 73 | ||
| BC014804.1 | - | 5 - 6 | 844 | 4489193 | 73 | |
| U39071.1 | - | 5 - 6 | 877 | 73 | ||
| U43508.1 | - | 5 - 6 | 839 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rorc
Genomic Map