Assay Details
Gene Symbol: Acsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027976.2 | NP_082252.1 | 16 - 17 | 2305 | 97 | |
GenBank mRNA | AK006541.1 | - | 5 - 6 | 782 | 97 | |
AK145894.1 | - | 16 - 17 | 1966 | 97 | ||
AK150542.1 | - | 16 - 17 | 1971 | 97 | ||
AK150663.1 | - | 16 - 17 | 1971 | 97 | ||
AK151378.1 | - | 16 - 17 | 1760 | 97 | ||
AK161024.1 | - | 16 - 17 | 1658 | 97 | ||
AK168019.1 | - | 4 - 5 | 306 | 97 | ||
AK178323.1 | - | 4 - 5 | 303 | 97 | ||
BC031544.1 | - | 16 - 17 | 1739 | 3987201 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
