Assay Details
Gene Symbol: Acsl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027976.2 | NP_082252.1 | 8 - 9 | 1573 | 75 | |
GenBank mRNA | AK034942.1 | - | 1573 | 75 | ||
AK145894.1 | - | 8 - 9 | 1234 | 75 | ||
AK150542.1 | - | 8 - 9 | 1239 | 75 | ||
AK150663.1 | - | 8 - 9 | 1239 | 75 | ||
AK151378.1 | - | 8 - 9 | 1028 | 75 | ||
AK161024.1 | - | 8 - 9 | 926 | 75 | ||
BC031544.1 | - | 8 - 9 | 1007 | 3987201 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
