Assay Details
| Gene Symbol: Xpnpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133216.3 | NP_573479.3 | 5 - 6 | 524 | 65 | |
| GenBank mRNA | AF363970.1 | - | 5 - 6 | 464 | 65 | |
| AK146033.1 | - | 5 - 6 | 504 | 65 | ||
| AK149676.1 | - | 5 - 6 | 463 | 65 | ||
| AK158597.1 | - | 3 - 4 | 482 | 65 | ||
| AK166715.1 | - | 5 - 6 | 424 | 65 | ||
| AK169024.1 | - | 4 - 5 | 350 | 65 | ||
| BC065174.1 | - | 5 - 6 | 412 | 6415314 | 65 | |
More Information
Set Membership: |
|

Xpnpep1
Genomic Map