Assay Details
| Gene Symbol: Sfxn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053198.3 | NP_444428.3 | 2 - 3 | 196 | 112 | |
| GenBank mRNA | AK148530.1 | - | 2 - 3 | 191 | 112 | |
| BC058211.1 | - | 2 - 3 | 193 | 112 | ||
| BC089535.1 | - | 2 - 3 | 178 | 6589805 | 112 | |
More Information
Set Membership: |
|

Sfxn4
Genomic Map