Assay Details
| Gene Symbol: Sfxn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053198.3 | NP_444428.3 | 8 - 9 | 496 | 72 | |
| GenBank mRNA | AF325263.1 | - | 8 - 9 | 438 | 72 | |
| AK148530.1 | - | 7 - 8 | 465 | 72 | ||
| BC058211.1 | - | 9 - 10 | 688 | 72 | ||
| BC089535.1 | - | 8 - 9 | 478 | 6589805 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sfxn4
Genomic Map