Assay Details
| Gene Symbol: Aldh7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127338.1 | NP_001120810.1 | 16 - 17 | 1585 | 126 | |
| NM_138600.4 | NP_613066.2 | 14 - 15 | 1391 | 126 | ||
| GenBank mRNA | AK004991.1 | - | 16 - 17 | 1519 | 126 | |
| AK145873.1 | - | 14 - 15 | 1334 | 126 | ||
| AK169195.1 | - | 14 - 15 | 1334 | 126 | ||
| AK198202.1 | - | 1 - 2 | 94 | 126 | ||
| BC012407.1 | - | 14 - 15 | 1353 | 4191228 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh7a1
Genomic Map