Assay Details
| Gene Symbol: Aldh7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127338.1 | NP_001120810.1 | 17 - 18 | 1685 | 135 | |
| NM_138600.4 | NP_613066.2 | 15 - 16 | 1491 | 135 | ||
| GenBank mRNA | AK004991.1 | - | 17 - 18 | 1619 | 135 | |
| AK145873.1 | - | 15 - 16 | 1434 | 135 | ||
| AK160117.1 | - | 15 - 16 | 1418 | 135 | ||
| AK169195.1 | - | 15 - 16 | 1434 | 135 | ||
| BC012407.1 | - | 15 - 16 | 1453 | 4191228 | 135 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aldh7a1
Genomic Map